Få mere at vide
2-Methyl-d3-propionitrile-3,3,3-d3, CDN
Artikelnummer. 30317939
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30317939 0.5 g 0.5g
30323797 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317939 Leverandør CDN Leverandørnr. CDND75190.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3,3,3-trideuterio-2-(trideuteriomethyl)propanenitrile
CAS 174736-88-0
Molekylær formel C4 2H6 H N
Mængde 0.5 g
InChI formel InChI=1 S/C4H7N/c1-4(2)3-5/h4H,1-2H3/i1D3,2D3
SMIL [2 H]C([2 H])([2 H])C(C#N)C([2 H])([2 H])[2 H]
IUPAC navn 3,3,3-trideuterio-2-(trideuteriomethyl)propanenitrile
Molekylvægt (g/mol) 75.142
Formel vægt 75.096 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity