Få mere at vide
(±)-2-Methyl-d3-butyric Acid, CDN
Artikelnummer. 30324241
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30324241 0.25 g -
30316275 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324241 Leverandør CDN Leverandørnr. CDND75350.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(trideuteriomethyl)butanoic acid
CAS 95926-92-4
Molekylær formel C5 2H3 H7 O2
Mængde 0.25 g
InChI formel InChI=1 S/C5H10O2/c1-3-4(2)5(6)7/h4H,3H2,1-2H3,(H,6,7)/i2D3
SMIL [2 H]C([2 H])([2 H])C(CC)C(=O)O
IUPAC navn 2-(trideuteriomethyl)butanoic acid
Molekylvægt (g/mol) 105.15
Formel vægt 105.087 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity