Få mere at vide
2-(Methyl-d3)-butanoic-2,3,3,4,4,4-d6 Acid, TRC
Artikelnummer. 30473714
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30473714 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473714 Leverandør Toronto Research Chemicals Leverandørnr. TRCM294866250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(Methyl-d3)-butanoic-2,3,3,4,4,4-d6 Acid
CAS 352431-44-8
Molekylær formel C5 D9 H O2
Mængde 250 mg
Synonym 2-Methylbutyric-d9 Acid, α-Methylbutyric-d9 Acid, (±)-2-Methylbutanoic-d9 Acid, (±)-2-Methylbutyric-d9 Acid, (±)-Methylethylacetic-d9 Acid, (±)-α-Methylbutyric-d9 Acid, 2-Ethylpropanoic-d9 Acid, 2-Methyl-n-butyric-d9 Acid, 2-Methylbutanoic-d9 Acid, 2-Methylbutyric-d9 Acid, Active Valeric-d9 Acid, DL-2-Methylbutanoic-d9 Acid, DL-2-Methylbutyric-d9 Acid, DL-α-Methylbutyric-d9 Acid, Ethylmethylacetic-d9 Acid, Methylethylacetic-d9 Acid, NSC 7304-d3, α-Methyl-n-butyric-d9 Acid
InChI formel InChI=1S/C5H10O2/c1-3-4(2)5(6)7/h4H,3H2,1-2H3,(H,6,7)/i1D3,2D3,3D2,4D
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])(C(=O)O)C([2H])([2H])[2H]
IUPAC navn 2,3,3,4,4,4-hexadeuterio-2-(trideuteriomethyl)butanoic acid
Molekylvægt (g/mol) 111.19
Formel vægt 111.1246