Få mere at vide
2-((Methyl-d3)amino)ethanol, TRC
Artikelnummer. 30479372
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30479372 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30479372 Leverandør Toronto Research Chemicals Leverandørnr. TRCM286092250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-((Methyl-d3)amino)ethanol
CAS 1216441-01-8
Molekylær formel C3 D3 H6 N O
Mængde 250 mg
Synonym (2-Hydroxyethyl)methylamine-d3, N-(Methyl-d3)-2-ethanolamine, Monomethylmonoethanolamine-d3, Methylethanolamine-d3, NSC 62776-d3
InChI formel InChI=1S/C3H9NO/c1-4-2-3-5/h4-5H,2-3H2,1H3/i1D3
SMIL [2H]C([2H])([2H])NCCO
IUPAC navn 2-(trideuteriomethylamino)ethanol
Molekylvægt (g/mol) 78.128
Formel vægt 78.087