Få mere at vide
2-Methyl-d3-3-(trifluoromethyl)pivalanilide, TRC
Artikelnummer. 30465095
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30465095 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30465095 Leverandør Toronto Research Chemicals Leverandørnr. TRCM330927250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methyl-d3-3-(trifluoromethyl)pivalanilide
CAS 1189715-10-3
Molekylær formel C13H13D3F3NO
Mængde 250 mg
Synonym 2,2-Dimethyl-N-[2-(methyl-d3)-3-(trifluoromethyl)phenyl] Propanamide
InChI formel InChI=1S/C13H16F3NO/c1-8-9(13(14,15)16)6-5-7-10(8)17-11(18)12(2,3)4/h5-7H,1-4H3,(H,17,18)/i1D3
SMIL O=C(C(C)(C)C)NC1=C(C([2H])([2H])[2H])C(C(F)(F)F)=CC=C1
Molekylvægt (g/mol) 262.29