Få mere at vide
2-Methyl-d3-1,4-naphthoquinone, CDN
Artikelnummer. 30322008
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322008 0.05 g
30321002 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322008 Leverandør CDN Leverandørnr. CDND51160.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Menadione-D3 (methyl-D3)
CAS 5172-16-7
Molekylær formel C11 D3 H5 O2
Mængde 0.05 g
Synonym 1,4-Naphthalenedione, 2-(methyl-d3)- (9 CI), 1,4-Naphthoquinone, 2-methyl-d3- (7 CI, 8 CI), 2-(Methyl-d3)-1,4-naphthalenedione, Menadione-d3, Menadione-D3 (methyl-D3), Vitamin K3-d3, 2-Methyl-d3-1,4-naphthoquinone
InChI formel InChI=1 S/C11H8O2/c1-7-6-10(12)8-4-2-3-5-9(8)11(7)13/h2-6 H,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])C1=CC(=O)c2ccccc2C1=O
IUPAC navn 2-(trideuteriomethyl)naphthalene-1,4-dione
Molekylvægt (g/mol) 175.1985
Formel vægt 175.0713 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity