Få mere at vide
2-Methyl-6-nitroaniline-d3, TRC
Artikelnummer. 30478424
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30478424 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30478424 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3235621MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methyl-6-nitroaniline-d3
CAS 1246816-42-1
Molekylær formel C7 D3 H5 N2 O2
Mængde 1 mg
Synonym 2-Methyl-6-nitro-benzenamine-d3, 6-Nitro-o-toluidine-d3, 1-Amino-2-methyl-6-nitrobenzene-d3, 1-Nitro-2-amino-3-methylbenzene-d3, 2-Amino-3-nitrotoluene-d3, 2-Methyl-6-nitroaniline-d3, 2-Nitro-6-methylaniline-d3, 3-Nitro-2-aminotoluene-d3, NSC 286-d3, NSC 52218-d3
InChI formel InChI=1S/C7H8N2O2/c1-5-3-2-4-6(7(5)8)9(10)11/h2-4H,8H2,1H3/i1D3
SMIL [2H]C([2H])([2H])c1cccc(c1N)[N+](=O)[O-]
IUPAC navn 2-nitro-6-(trideuteriomethyl)aniline
Molekylvægt (g/mol) 155.169
Formel vægt 155.077