Learn More
2-Methyl-4-pentenoic Acid, TRC
Product Code. 30473728
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity Unit Size
30473728 500 mg 500mg
1 options
Product Code. 30473728 Supplier Toronto Research Chemicals Supplier No. TRCM220778500MG
 Restricted Item
Forventet forsendelse: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Methyl-4-pentenoic Acid
CAS 1575-74-2
Molecular Formula C6H10O2
Quantity 500 mg
InChI Formula InChI=1S/C6H10O2/c1-3-4-5(2)6(7)8/h3,5H,1,4H2,2H3,(H,7,8)
SMILES CC(CC=C)C(=O)O
IUPAC Name 2-methylpent-4-enoic acid
Molecular Weight (g/mol) 114.14
Formula Weight 114.0681