Få mere at vide
2-Methyl-4-nitroaniline-d3, TRC
Artikelnummer. 30467695
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30467695 10 mg
30465249 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30467695 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32354710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1246815-44-0
Molekylær formel C7H5D3N2O2
Molekylvægt (g/mol) 155.17
Synonym 2-Methyl-4-nitro-benzenamine-d3, 4-Nitro-o-toluidine-d3, 1-Amino-2-methyl-4-nitrobenzene-d3, 2-Amino-1-methyl-5-nitrobenzene-d3, 2-Amino-5-nitrotoluene-d3, 2-Methyl-4-nitrobenzenamine-d3, 2-Methyl-4-nitrophenylamine-d3, Fast Red RL Base-d3
SMIL [2H]C([2H])([2H])C1=C(C=CC(=C1)[N+](=O)[O-])N
Tekniske data
Kemisk navn eller materiale 2-Methyl-4-nitroaniline-d3
Mængde 10 mg
InChI formel InChI=1S/C7H8N2O2/c1-5-4-6(9(10)11)2-3-7(5)8/h2-4H,8H2,1H3/i1D3
Formel vægt 155.0774