Få mere at vide
2-Methyl-4,6-dinitrophenol, TRC
Artikelnummer. 30479643
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479643 500 mg
30478552 100 mg
30478553 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30479643 Leverandør Toronto Research Chemicals Leverandørnr. TRCM221976500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 534-52-1
Molekylær formel C7 H6 N2 O5
Molekylvægt (g/mol) 198.13
Synonym Phenol, 2-methyl-4,6-dinitro-, o-Cresol, 4,6-dinitro- (8CI), 2-Methyl-4,6-dinitrophenol, 2,4-Dinitro-6-methylphenol, 3,5-Dinitro-2-hydroxytoluene, 4,6-Dinitro-2-methylphenol, 4,6-Dinitro-o-cresol, 6-Methyl-2,4-dinitrophenol, Arborol, DNOC, Degrassan, Dekrysil, Detal, Dillex, Dinitro, Dinitro-o-cresol, Dinitrocresol, Dinitrodendtroxal, Dinitrol, Dinoc, Dinurania, Ditrosol, Effusan, Effusan 3436, Elgetol, Elgetol 30, Elipol, Extrar, Flavin-Sandoz, Hedolit, Hedolite, K III, K IV, Kreozan, Krezotol 50, Lipan, NSC 2082, Neudorff DN 50, Nitrofan, Prokarbol, Rafex, Rafex 35, Raphatox, Sandolin, Sandolin A, Selinon, Sinox, Winterwash
IUPAC navn 2-methyl-4,6-dinitrophenol
SMIL Cc1cc(cc(c1O)[N+](=O)[O-])[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 2-Methyl-4,6-dinitrophenol
Mængde 500 mg
InChI formel InChI=1S/C7H6N2O5/c1-4-2-5(8(11)12)3-6(7(4)10)9(13)14/h2-3,10H,1H3
Formel vægt 198.0277