Få mere at vide
2-Methyl-4,6-dinitrophenol 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-Methyl-4,6-dinitrophenol |
| CAS | 534-52-1 |
| Analyt- eller komponentnavne | 2-Methyl-4,6-dinitrophenol |
| Forsendelsestilstand | +20°C |
| Molekylær formel | C7 H6 N2 O5 |
| Koncentration | 1000 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Phenol, 2-methyl-4,6-dinitro-, o-Cresol, 4,6-dinitro- (8CI), 2-Methyl-4,6-dinitrophenol, 2,4-Dinitro-6-methylphenol, 3,5-Dinitro-2-hydroxytoluene, 4,6-Dinitro-2-methylphenol, 4,6-Dinitro-o-cresol, 6-Methyl-2,4-dinitrophenol, Arborol, DNOC, Degrassan, Dekrysil, Detal, Dillex, Dinitro, Dinitro-o-cresol, Dinitrocresol, Dinitrodendtroxal, Dinitrol, Dinoc, Dinurania, Ditrosol, Effusan, Effusan 3436, Elgetol, Elgetol 30, Elipol, Extrar, Flavin-Sandoz, Hedolit, Hedolite, K III, K IV, Kreozan, Krezotol 50, Lipan, NSC 2082, Neudorff DN 50, Nitrofan, Prokarbol, Rafex, Rafex 35, Raphatox, Sandolin, Sandolin A, Selinon, Sinox, Winterwash |
| InChI formel | InChI=1S/C7H6N2O5/c1-4-2-5(8(11)12)3-6(7(4)10)9(13)14/h2-3,10H,1H3 |
| SMIL | Cc1cc(cc(c1O)[N+](=O)[O-])[N+](=O)[O-] |
| Vis mere |