missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-(trifluoromethyl)benzonitrile, 97%, Thermo Scientific™

Catalog Number 11495940
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
261952-02-7
C9H6F3N
185.15
MFCD01631593
HXGUSYYIWFJFGY-UHFFFAOYSA-N
2-methyl-3-trifluoromethyl benzonitrile, 3-cyano-2-methylbenzotrifluoride, 2-cyano-6-trifluoromethyl toluene, 2-methyl trifluoromethyl benzonitrile, 2-methyl-3-trifluoromethyl benzenecarbonitrile
2778934
2-methyl-3-(trifluoromethyl)benzonitrile
CC1=C(C=CC=C1C(F)(F)F)C#N
This item is not returnable. View return policy
261952-02-7
C9H6F3N
185.15
MFCD01631593
HXGUSYYIWFJFGY-UHFFFAOYSA-N
2-methyl-3-trifluoromethyl benzonitrile, 3-cyano-2-methylbenzotrifluoride, 2-cyano-6-trifluoromethyl toluene, 2-methyl trifluoromethyl benzonitrile, 2-methyl-3-trifluoromethyl benzenecarbonitrile
2778934
2-methyl-3-(trifluoromethyl)benzonitrile
CC1=C(C=CC=C1C(F)(F)F)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 28°C to 31°C
Quantity 1 g
UN Number UN3439
Formula Weight 185.15
Percent Purity 97%
Chemical Name or Material 2-Methyl-3-(trifluoromethyl)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'