Få mere at vide
2-Methyl-3-phenylbenzoic Acid-d5 Methyl Ester, TRC
Artikelnummer. 30469705
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30469705 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469705 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32069250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methyl-3-phenylbenzoic Acid-d5 Methyl Ester
CAS 1329835-46-2
Molekylær formel C15H9D5O2
Mængde 50 mg
Synonym 2-Methyl-[1,1'-biphenyl]-3-carboxylic Acid-d5 Methyl Ester
InChI formel InChI=1S/C15H14O2/c1-11-13(12-7-4-3-5-8-12)9-6-10-14(11)15(16)17-2/h3-10H,1-2H3/i3D,4D,5D,7D,8D
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2cccc(C(=O)OC)c2C
IUPAC navn methyl 2-methyl-3-(2,3,4,5,6-pentadeuteriophenyl)benzoate
Molekylvægt (g/mol) 231.3
Formel vægt 231.1308