Få mere at vide
2-Methyl-3-biphenylmethanol-d5, TRC
Artikelnummer. 30472659
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30472659 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472659 Leverandør Toronto Research Chemicals Leverandørnr. TRCM29413210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methyl-3-biphenylmethanol-d5
CAS 1329835-20-2
Molekylær formel C14H9D5O
Mængde 10 mg
Synonym 2-Methyl-[1,1'-biphenyl]-3-methanol-d5, 2-Methyl-3-phenylbenzyl-d5 Alcohol, 2-Methylbiphenyl-3-ylmethanol-d5, 3-Hydroxymethyl-2-methylbiphenyl-d5
InChI formel InChI=1S/C14H14O/c1-11-13(10-15)8-5-9-14(11)12-6-3-2-4-7-12/h2-9,15H,10H2,1H3/i2D,3D,4D,6D,7D
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2cccc(CO)c2C
IUPAC navn [2-methyl-3-(2,3,4,5,6-pentadeuteriophenyl)phenyl]methanol
Molekylvægt (g/mol) 203.29
Formel vægt 203.1358