Få mere at vide
2-Methyl-2-[(trimethylsilyl)oxy]propanenitrile-d6, TRC
Artikelnummer. 30335646
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30335646 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30335646 Leverandør Toronto Research Chemicals Leverandørnr. TRCB488282100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methyl-2-[(trimethylsilyl)oxy]propanenitrile-d6
CAS 107090-98-2
Molekylær formel C7H9D6NOSi
Mængde 100 mg
InChI formel InChI=1S/C7H15NOSi/c1-7(2,6-8)9-10(3,4)5/h1-5H3/i1D3,2D3
SMIL [2H]C([2H])([2H])C(O[Si](C)(C)C)(C#N)C([2H])([2H])[2H]
IUPAC navn 3,3,3-trideuterio-2-(trideuteriomethyl)-2-trimethylsilyloxypropanenitrile
Molekylvægt (g/mol) 163.32