Få mere at vide
2-Methyl-2-propyl-1,3-propanediol, TRC
Artikelnummer. 30479267
Skift visning
Mængde:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479267 50 g
30476281 25 g
30466168 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30479267 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32059050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 78-26-2
Molekylær formel C7 H16 O2
Molekylvægt (g/mol) 132.2
Synonym 2-Methyl-2-propylpropane-1,3-diol, 1,3-Propanediol, 2-methyl-2-propyl-, 2-Methyl-2-propyl-1,3-propanediol, 2-Propyl-2-methyl-1,3-propanediol, NSC 26233
IUPAC navn 2-methyl-2-propylpropane-1,3-diol
SMIL CCCC(C)(CO)CO
Tekniske data
Kemisk navn eller materiale 2-Methyl-2-propylpropane-1,3-diol
Mængde 50 g
InChI formel InChI=1S/C7H16O2/c1-3-4-7(2,5-8)6-9/h8-9H,3-6H2,1-2H3
Formel vægt 132.115