missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Methyl-2-propoxypropane, TRC

Artikelnummer. 30464571
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30464571 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30464571 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3293061G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Methyl-2-propoxypropane
CAS 29072-93-3
Molekylær formel C7H16O
Mængde 1 g
Synonym 2-Methyl-2-propoxypropane, 1,1-Dimethylethyl Propyl Ether, 1-tert-Butoxypropane, Propyl 1,1-Dimethylethyl Ether, Propyl tert-Butyl Ether, n-Propyl tert-Butyl Ether, tert-Butyl n-Propyl Ether, tert-Butyl Propyl Ether
InChI formel InChI=1S/C7H16O/c1-5-6-8-7(2,3)4/h5-6H2,1-4H3
SMIL CCCOC(C)(C)C
IUPAC navn 2-methyl-2-propoxypropane
Molekylvægt (g/mol) 116.2
Formel vægt 116.1201
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.