Få mere at vide
2-Methyl-1H-quinolin-4-one, TRC
Artikelnummer. 30008692
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008692 100 mg
30006170 500 mg
30008859 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008692 Leverandør Toronto Research Chemicals Leverandørnr. TRCM396080100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5660-24-2
Molekylær formel C10H9NO
Molekylvægt (g/mol) 159.18
Synonym 2-Methyl-4(1H)-quinolinone, 2-Methyl-1,4-dihydro-4-oxoquinoline, 2-Methyl-1,4-dihydroquinolin-4-one, 2-Methyl-1H-quinolin-4-one, 2-Methyl-4(1H)-quinolone, 2-Methyl-4-quinolinone, 4(1H)-Quinaldone
IUPAC navn 2-methyl-1H-quinolin-4-one
SMIL CC1=CC(=O)c2ccccc2N1
Tekniske data
Kemisk navn eller materiale 2-Methyl-1H-quinolin-4-one
Mængde 100 mg
InChI formel InChI=1S/C10H9NO/c1-7-6-10(12)8-4-2-3-5-9(8)11-7/h2-6H,1H3,(H,11,12)
Formel vægt 159.0684