Få mere at vide
2-Methyl-1-phenylpropane-d14, CDN
Artikelnummer. 30326902
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326902 1 g
30321000 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326902 Leverandør CDN Leverandørnr. CDND50491G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isobutylbenzene-D14
CAS 350818-58-5
Molekylær formel C10 D14
Mængde 1 g
Synonym Benzene-d5, [2-(methyl-d3)propyl-1,1,2,3,3,3-d6]- (9 CI), [2-(Methyl-d3)propyl-1,1,2,3,3,3-d5]benzene-2,3,4,5,6-d5, 2-(Methyl-d3)-1-(phenyl-d5)propane-1,1,2,3,3,3-d5, Isobutylbenzene-d14, Isobutylbenzene-D14
InChI formel InChI=1 S/C10H14/c1-9(2)8-10-6-4-3-5-7-10/h3-7,9 H,8H2,1-2H3/i1D3,2D3,3 D,4 D,5 D,6 D,7 D,8D2,9 D
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-[1,1,2,3,3,3-hexadeuterio-2-(trideuteriomethyl)propyl]benzene
Molekylvægt (g/mol) 148.3044
Formel vægt 148.1974 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity