Få mere at vide
2-Methyl-1,4-naphthalenediol-d8, TRC
Artikelnummer. 30465760
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30465760 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30465760 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3231371MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methyl-1,4-naphthalenediol-d8
CAS 1142409-65-1
Molekylær formel C11H2D8O2
Mængde 1 mg
Synonym 1,4-Dihydroxy-2-methylnaphthalene-d8, 2-Methyl-1,4-dihydroxynaphthalene-d8, 2-Methyl-1,4-naphthohydroquinone-d8, 2-Methyl-1,4-naphthoquinol-d8, 2-Methylhydronaphthoquinone-d8, 2-Methylnaphthalene-1,4-diol-d8, 4-Hydroxy-3-methyl-alpha-naphthol-d8, Dihydrovitamin K3-d8, Menadiol-d8, Reduced Menadione-d8, Reduced Vitamin K3-d8, Vitamin K3H2-d8
InChI formel InChI=1S/C11H10O2/c1-7-6-10(12)8-4-2-3-5-9(8)11(7)13/h2-6,12-13H,1H3/i1D3,2D,3D,4D,5D,6D
SMIL [2H]c1c([2H])c([2H])c2c(O)c(c([2H])c(O)c2c1[2H])C([2H])([2H])[2H]
IUPAC navn 2,5,6,7,8-pentadeuterio-3-(trideuteriomethyl)naphthalene-1,4-diol
Molekylvægt (g/mol) 182.25
Formel vægt 182.1183