Få mere at vide
2-Methyl-1,3-dioxolane-2-propanol-d4, TRC
Artikelnummer. 30472552
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30472552 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472552 Leverandør Toronto Research Chemicals Leverandørnr. TRCM303627250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methyl-1,3-dioxolane-2-propanol-d4
CAS 1215630-77-5
Molekylær formel C7H10D4O3
Mængde 250 mg
Synonym 4,4-(Ethylenedioxy)-1-pentanol
InChI formel InChI=1S/C7H14O3/c1-7(3-2-4-8)9-5-6-10-7/h8H,2-6H2,1H3/i2D2,4D2
SMIL [2H]C([2H])(O)C([2H])([2H])CC1(C)OCCO1
IUPAC navn 1,1,2,2-tetradeuterio-3-(2-methyl-1,3-dioxolan-2-yl)propan-1-ol
Molekylvægt (g/mol) 150.21
Formel vægt 150.1194