missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Methyl-1,3-dioxolane-2-propanoic Acid Methyl Ester-d2, TRC

Artikelnummer. 30473476
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473476 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473476 Leverandør Toronto Research Chemicals Leverandørnr. TRCM30366225MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2-Methyl-1,3-dioxolane-2-propanoic Acid Methyl Ester-d2
CAS 1216512-70-7
Molekylær formel C8H12D2O4
Mængde 25 mg
Synonym Methyl 2-Methyl-1,3-dioxolane-2-propionate-d2, Methyl Levulinate Ethylene Ketal-d2, Methyl 3-(2-Methyl-1,3-dioxolan-2-yl)propanoate-d2
InChI formel InChI=1S/C8H14O4/c1-8(11-5-6-12-8)4-3-7(9)10-2/h3-6H2,1-2H3/i3D2
SMIL [2H]C([2H])(CC1(C)OCCO1)C(=O)OC
IUPAC navn methyl 2,2-dideuterio-3-(2-methyl-1,3-dioxolan-2-yl)propanoate
Molekylvægt (g/mol) 176.21
Formel vægt 176.1018
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.