missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-benzothiazol-5-amine, 95%, ACROS Organics™

Brand:  Acros Organics 432760050

Additional Details : CAS Number : 13382-43-9 Weight : 0.05500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10452498

  • 960.00 DKK / 5g

Estimated Shipment
10-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

13382-43-9
164.23
GPWQHYMVUZYWIK-UHFFFAOYSA-N
36229
CC1=NC2=C(S1)C=CC(=C2)N
C8H8N2S
MFCD00226291
2-methylbenzo d thiazol-5-amine, 5-amino-2-methylbenzothiazole, 5-benzothiazolamine, 2-methyl, 2-methyl-5-benzothiazolamine, 2-methyl-5-aminobenzothiazole, 2-methylbenzothiazole-5-ylamine, 2-methyl-1,3-benzothiazol-5-ylamine, acmc-209v5w, cambridge id 5107842, 2-methylbenzothiazol-5-amine
2-methyl-1,3-benzothiazol-5-amine

Specifications

2-Methyl-1, 3-benzothiazol-5-amine
101°C to 103.5°C
13382-43-9
100.0
C8H8N2S
2-methylbenzo d thiazol-5-amine, 5-amino-2-methylbenzothiazole, 5-benzothiazolamine, 2-methyl, 2-methyl-5-benzothiazolamine, 2-methyl-5-aminobenzothiazole, 2-methylbenzothiazole-5-ylamine, 2-methyl-1,3-benzothiazol-5-ylamine, acmc-209v5w, cambridge id 5107842, 2-methylbenzothiazol-5-amine
CC1=NC2=C(S1)C=CC(=C2)N
164.23
164.23
Glass bottle
5g
94.0
94% min. (HPLC)
MFCD00226291
GPWQHYMVUZYWIK-UHFFFAOYSA-N
2-methyl-1,3-benzothiazol-5-amine
36229
95%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Methyl-1,3-benzothiazol-5-amine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents