missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxythiazole, 98%, Thermo Scientific™

Catalog Number 15463297
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
14542-13-3
C4H5NOS
115.15
MFCD01631143
MJJRDTKNLLMJDJ-UHFFFAOYSA-N
2-methoxythiazole, thiazole, 2-methoxy, 2-methoxy thiazole, acmc-209cuy, #, ksc174c3j, 2-methoxy-1,3-thiazole
575451
2-methoxy-1,3-thiazole
COC1=NC=CS1
This item is not returnable. View return policy
14542-13-3
C4H5NOS
115.15
MFCD01631143
MJJRDTKNLLMJDJ-UHFFFAOYSA-N
2-methoxythiazole, thiazole, 2-methoxy, 2-methoxy thiazole, acmc-209cuy, #, ksc174c3j, 2-methoxy-1,3-thiazole
575451
2-methoxy-1,3-thiazole
COC1=NC=CS1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 150°C to 151°C
Flash Point 49°C (120°F)
Quantity 5 g
UN Number UN1993
Formula Weight 115.15
Percent Purity 98%
Chemical Name or Material 2-Methoxythiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'