Få mere at vide
2-Methoxytetrahydrofuran, TRC
Artikelnummer. 30466755
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30466755 10 g
30481215 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30466755 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26450510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13436-45-8
Molekylær formel C5H10O2
Molekylvægt (g/mol) 102.13
Synonym Tetrahydro-2-methoxyfuran
IUPAC navn 2-methoxyoxolane
SMIL COC1CCCO1
Tekniske data
Kemisk navn eller materiale 2-Methoxytetrahydrofuran
Mængde 10 g
InChI formel InChI=1S/C5H10O2/c1-6-5-3-2-4-7-5/h5H,2-4H2,1H3
Formel vægt 102.0681