Få mere at vide
2-Methoxyphenol-3,4,5,6-d4, TRC
Artikelnummer. 30468774
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30468774 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30468774 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26530110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methoxyphenol-3,4,5,6-d4
CAS 7329-52-4
Molekylær formel C7 D4 H4 O2
Mængde 10 mg
Synonym Phen-2,3,4,5-d4-ol','6-methoxy- (7CI','8CI','9CI','ACI), 6-Methoxyphen-2,3,4,5-d4-ol (ACI), 3,4,5,6-Tetradeuteroguaiacol, 2-Methoxyphenol-3,4,5,6-d4, Guaiacol-d4 (ring-d4), 2-Methoxyphenol-3,4,5,6-D4, 2-Methoxyphenol-d4
InChI formel InChI=1S/C7H8O2/c1-9-7-5-3-2-4-6(7)8/h2-5,8H,1H3/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(OC)c(O)c1[2H]
IUPAC navn 2,3,4,5-tetradeuterio-6-methoxyphenol
Molekylvægt (g/mol) 128.1619
Formel vægt 128.0775