Få mere at vide
2-Methoxyphenol-3,4,5,6-d4,OD, CDN
Artikelnummer. 30317657
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30317657 0.5 g 0.5g
30314289 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317657 Leverandør CDN Leverandørnr. CDND61840.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methoxyphenol-O,3,4,5,6-D5
CAS 20189-11-1
Molekylær formel C7 D5 H3 O2
Mængde 0.5 g
Synonym Phen-2,3,4,5-d4-ol-d, 6-methoxy- (8 CI, 9 CI), 2-Methoxyphen-d4-ol-d, OD-3,4,5,6-Tetradeuteroguaiacol, 2-Methoxyphenol-O,3,4,5,6-d5, Guaiacol-d4 (OD,ring-d4), 2-Methoxyphenol-O,3,4,5,6-D5, 2-Methoxyphenol-d5, 2-Methoxyphenol-3,4,5,6-d4,OD
InChI formel InChI=1 S/C7H8O2/c1-9-7-5-3-2-4-6(7)8/h2-5,8 H,1H3/i2D,3 D,4 D,5 D/hD
SMIL [2 H]Oc1c([2 H])c([2 H])c([2 H])c([2 H])c1OC
IUPAC navn 1,2,3,4-tetradeuterio-5-deuteriooxy-6-methoxybenzene
Molekylvægt (g/mol) 129.168
Formel vægt 129.0838 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity