Få mere at vide
2-Methoxyphenol-3,4,5,6-d4, CDN
Artikelnummer. 30314703
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30314703 0.5 g 0.5g
30309104 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314703 Leverandør CDN Leverandørnr. CDND61530.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methoxyphenol-3,4,5,6-d4
CAS 7329-52-4
Molekylær formel C7 D4 H4 O2
Mængde 0.5 g
Synonym Phen-2,3,4,5-d4-ol, 6-methoxy- (7 CI, 8 CI, 9 CI, ACI), 6-Methoxyphen-2,3,4,5-d4-ol (ACI), 3,4,5,6-Tetradeuteroguaiacol, 2-Methoxyphenol-3,4,5,6-d4, Guaiacol-d4 (ring-d4), 2-Methoxyphenol-3,4,5,6-D4, 2-Methoxyphenol-d4
InChI formel InChI=1 S/C7H8O2/c1-9-7-5-3-2-4-6(7)8/h2-5,8 H,1H3/i2D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])c(OC)c(O)c1[2 H]
IUPAC navn 2,3,4,5-tetradeuterio-6-methoxyphenol
Molekylvægt (g/mol) 128.1619
Formel vægt 128.0775 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity