Få mere at vide
2-Methoxynaphthalen, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-Methoxynaphthalene(Nerolin) |
| CAS | 93-04-9 |
| Molekylær formel | C11 H10 O |
| Mængde | 5 g |
| Synonym | 2-Methoxynaphthalene(Nerolin), Naproxen Imp. M (EP), Naphthalene','2-methoxy- (8CI','9CI','ACI), 2-Methoxynaphthalene (ACI), Ether','methyl 2-naphthyl (3CI), 2-Naphthol methyl ether, 2-Naphthyl methyl ether, 6-Methoxy-2-naphthalene, Methyl 2-naphthyl ether, Methyl β-naphthyl ether, Nerolin, Nerolin (old), Nerolin yara yara, NSC 4171, Yara yara, β-Methoxynaphthalene, β-Naphthol methyl ether, β-Naphthyl methyl ether |
| InChI formel | InChI=1S/C11H10O/c1-12-11-7-6-9-4-2-3-5-10(9)8-11/h2-8H,1H3 |
| SMIL | COc1ccc2ccccc2c1 |
| IUPAC navn | 2-methoxynaphthalen |
| Molekylvægt (g/mol) | 158.2 |
| Formel vægt | 158.0732 |