Få mere at vide
2-Methoxyestrone-1,4,16,16-d4, TRC
Artikelnummer. 30471043
Skift visning
Mængde:
50 μg
Artikelnummer. Mængde
30471043 50 μg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30471043 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26252150UG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methoxyestrone-1,4,16,16-d4
CAS 949885-90-9
Molekylær formel C19 D4 H20 O3
Mængde 50 μg
Synonym Estra-1,3,5(10)-trien-17-one-1,4,16,16-d4, 3-hydroxy-2-methoxy-, 2-Methoxyestrone-(1,4,16,16-D4)
InChI formel InChI=1S/C19H24O3/c1-19-8-7-12-13(15(19)5-6-18(19)21)4-3-11-9-16(20)17(22-2)10-14(11)12/h9-10,12-13,15,20H,3-8H2,1-2H3/t12-,13+,15-,19-/m0/s1/i6D2,9D,10D
SMIL [2H]c1c(O)c(OC)c([2H])c2[C@H]3CC[C@@]4(C)[C@@H](CC([2H])([2H])C4=O)[C@@H]3CCc12
IUPAC navn (8R,9S,13S,14S)-1,4,16,16-tetradeuterio-3-hydroxy-2-methoxy-13-methyl-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthren-17-one
Molekylvægt (g/mol) 304.42
Formel vægt 304.1977