Få mere at vide
2-Methoxyestrone-1,4,16,16-d4, TRC
Artikelnummer. 30478928
Skift visning
Mængde:
0.1 mg
0.5 mg
Pakningsstørrelse:
0.1mg
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30478928 0.5 mg 0.5mg
30483271 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30478928 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2625210.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 949885-90-9
Molekylær formel C19 D4 H20 O3
Molekylvægt (g/mol) 304.42
Synonym Estra-1,3,5(10)-trien-17-one-1,4,16,16-d4, 3-hydroxy-2-methoxy-, 2-Methoxyestrone-(1,4,16,16-D4)
IUPAC navn (8R,9S,13S,14S)-1,4,16,16-tetradeuterio-3-hydroxy-2-methoxy-13-methyl-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthren-17-one
SMIL [2H]c1c(O)c(OC)c([2H])c2[C@H]3CC[C@@]4(C)[C@@H](CC([2H])([2H])C4=O)[C@@H]3CCc12
Tekniske data
Kemisk navn eller materiale 2-Methoxyestrone-1,4,16,16-d4
Mængde 0.5 mg
InChI formel InChI=1S/C19H24O3/c1-19-8-7-12-13(15(19)5-6-18(19)21)4-3-11-9-16(20)17(22-2)10-14(11)12/h9-10,12-13,15,20H,3-8H2,1-2H3/t12-,13+,15-,19-/m0/s1/i6D2,9D,10D
Formel vægt 304.1977