missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog Number p-7032830
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
10130-87-7
C7H7ClO3S
206.64
MFCD01961367
GYOBZOBUOMDRRN-UHFFFAOYSA-N
2-methoxy-benzenesulfonyl chloride, 2-methoxybenzene-1-sulfonyl chloride, 2-methoxybenzenesulfonylchloride, o-anisolesulfonyl chloride, 2-methoxyphenylsulfonyl chloride, acmc-1bntp, methoxyphenylsulfonyl chloride, 2-methoxybenzensulfonylchloride, 2-methoxybenzenesulphonylchloride, 2-methoxybenzene sulfonylchloride
3745851
2-methoxybenzenesulfonyl chloride
COC1=CC=CC=C1S(=O)(=O)Cl
This item is not returnable. View return policy
10130-87-7
C7H7ClO3S
206.64
MFCD01961367
GYOBZOBUOMDRRN-UHFFFAOYSA-N
2-methoxy-benzenesulfonyl chloride, 2-methoxybenzene-1-sulfonyl chloride, 2-methoxybenzenesulfonylchloride, o-anisolesulfonyl chloride, 2-methoxyphenylsulfonyl chloride, acmc-1bntp, methoxyphenylsulfonyl chloride, 2-methoxybenzensulfonylchloride, 2-methoxybenzenesulphonylchloride, 2-methoxybenzene sulfonylchloride
3745851
2-methoxybenzenesulfonyl chloride
COC1=CC=CC=C1S(=O)(=O)Cl

For synthesis and pharmacological evaluation of sulfonamide derivatives of thiazolidin-4-ones 5-Bromo-2-methoxybenzenesulfonyl chloride is used.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
For synthesis and pharmacological evaluation of sulfonamide derivatives of thiazolidin-4-ones 5-Bromo-2-methoxybenzenesulfonyl chloride is used.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
TRUSTED_SUSTAINABILITY
Melting Point 50°C to 54°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 206.65
Percent Purity 97%
Chemical Name or Material 2-Methoxybenzenesulfonyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'