Få mere at vide
2-Methoxyacetic Acid-d3, TRC
Artikelnummer. 30474108
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30474108 100 mg
30469802 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30474108 Leverandør Toronto Research Chemicals Leverandørnr. TRCM260827100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 345910-00-1
Molekylær formel C3 2H3 H3 O3
Molekylvægt (g/mol) 93.1
Synonym Acetic acid, methoxy-d3- (9CI), 2-(Methoxy-d3)acetic acid, Methoxyacetic acid-d3, (Methoxy-d3)ethanoic acid, Methoxy-d3-acetic acid, Trideuteriomethoxyacetic acid, 2-Methoxyacetic Acid D3 (methoxy-D3)
IUPAC navn 2-(trideuteriomethoxy)acetic acid
SMIL [2H]C([2H])([2H])OCC(=O)O
Tekniske data
Kemisk navn eller materiale 2-Methoxyacetic acid methoxy-d3
Mængde 100 mg
InChI formel InChI=1S/C3H6O3/c1-6-2-3(4)5/h2H2,1H3,(H,4,5)/i1D3
Formel vægt 93.0505