missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-N-methylbenzamide, 97%, Thermo Scientific™

Catalog Number 15453378
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
3400-35-9
C9H11NO2
165.19
MFCD01578285
NXVAEVHJWOBXKG-UHFFFAOYSA-N
benzamide, o-methoxy-n-methyl, o-methoxy-n-methylbenzamide, n-methyl-2-methoxybenzamide, 2-methoxyphenyl-n-methylcarboxamide
76940
2-methoxy-N-methylbenzamide
CNC(=O)C1=CC=CC=C1OC
This item is not returnable. View return policy
3400-35-9
C9H11NO2
165.19
MFCD01578285
NXVAEVHJWOBXKG-UHFFFAOYSA-N
benzamide, o-methoxy-n-methyl, o-methoxy-n-methylbenzamide, n-methyl-2-methoxybenzamide, 2-methoxyphenyl-n-methylcarboxamide
76940
2-methoxy-N-methylbenzamide
CNC(=O)C1=CC=CC=C1OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 2638881
Formula Weight 165.19
Percent Purity 97%
Chemical Name or Material 2-Methoxy-N-methylbenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'