Learn More
Alfa Aesar™ 2-Methoxy-N-methylbenzamide, 97%
Brand:  Alfa Aesar™ H56836.03
Additional Details : CAS Number : 3400-35-9 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15443378
  • 4457.00 DKK / 1g
Estimated Shipment
25-03-2021
Descripción
Descripción
Especificaciones
Identificadores de productos químicos
3400-35-9
165.192
NXVAEVHJWOBXKG-UHFFFAOYSA-N
76940
CNC(=O)C1=CC=CC=C1OC
C9H11NO2
MFCD01578285
benzamide, o-methoxy-n-methyl, o-methoxy-n-methylbenzamide, n-methyl-2-methoxybenzamide, 2-methoxyphenyl-n-methylcarboxamide
2-methoxy-N-methylbenzamide
Especificaciones
2-Methoxy-N-methylbenzamide
3400-35-9
MFCD01578285
benzamide, o-methoxy-n-methyl, o-methoxy-n-methylbenzamide, n-methyl-2-methoxybenzamide, 2-methoxyphenyl-n-methylcarboxamide
CNC(=O)C1=CC=CC=C1OC
165.192
165.19
1g
C9H11NO2
2638881
NXVAEVHJWOBXKG-UHFFFAOYSA-N
2-methoxy-N-methylbenzamide
76940
97%
Videos
SDS Documentos