Få mere at vide
2-Methoxy-d3-aniline, CDN
Artikelnummer. 30320461
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320461 1 g -
30319499 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320461 Leverandør CDN Leverandørnr. CDND73621G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale o-Anisidine methyl-d3
CAS 1398066-00-6
Molekylær formel C7 D3 H6 N O
Mængde 1 g
InChI formel InChI=1 S/C7H9NO/c1-9-7-5-3-2-4-6(7)8/h2-5 H,8H2,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])Oc1ccccc1N
IUPAC navn 2-(trideuteriomethoxy)aniline
Molekylvægt (g/mol) 126.17
Formel vægt 126.087 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity