Få mere at vide
2-(Methoxy-d3)-3-(1-methylethyl)pyrazine, TRC
Artikelnummer. 30476556
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30476556 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476556 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2643635MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(Methoxy-d3)-3-(1-methylethyl)pyrazine
CAS 588732-60-9
Molekylær formel C8 2H3 H9 N2 O
Mængde 5 mg
Synonym Pyrazine','2-(methoxy-d3)-3-(1-methylethyl)- (9CI), 2-Isopropyl-3-methoxy-d3-pyrazine, 2-(Methoxy-d3)-3-(1-methylethyl)pyrazine, 2-Isopropyl-3-(methoxy-d3)pyrazine, 2-Isopropyl-3-methoxypyrazine-d3, 2-Isopropyl-3-methoxypyrazine-D3 (methoxy-D3)
InChI formel InChI=1S/C8H12N2O/c1-6(2)7-8(11-3)10-5-4-9-7/h4-6H,1-3H3/i3D3
SMIL [2H]C([2H])([2H])Oc1nccnc1C(C)C
IUPAC navn 2-propan-2-yl-3-(trideuteriomethoxy)pyrazine
Molekylvægt (g/mol) 155.212
Formel vægt 155.114