missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-6-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

Catalog Number 15488205
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1017779-04-2
C9H8BrF3O
269.061
MFCD09832326
WJUBRZQZRVVXBG-UHFFFAOYSA-N
2-methoxy-6-trifluoromethyl benzyl bromide, 2-bromomethyl-1-methoxy-3-trifluoromethyl benzene, 2-methoxy-6-trifluoromethyl benzyl bromide, jrd
46737596
2-(bromomethyl)-1-methoxy-3-(trifluoromethyl)benzene
COC1=CC=CC(=C1CBr)C(F)(F)F
This item is not returnable. View return policy
1017779-04-2
C9H8BrF3O
269.061
MFCD09832326
WJUBRZQZRVVXBG-UHFFFAOYSA-N
2-methoxy-6-trifluoromethyl benzyl bromide, 2-bromomethyl-1-methoxy-3-trifluoromethyl benzene, 2-methoxy-6-trifluoromethyl benzyl bromide, jrd
46737596
2-(bromomethyl)-1-methoxy-3-(trifluoromethyl)benzene
COC1=CC=CC(=C1CBr)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 42°C to 45°C
Quantity 250 mg
UN Number UN3261
Formula Weight 269.06
Percent Purity 97%
Chemical Name or Material 2-Methoxy-6-(trifluoromethyl)benzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'