missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-6-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™

Catalog Number p-7031290
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
681510-01-0
C9H6ClF3O2
238.59
MFCD09832320
HXPBLTYRTHTMFQ-UHFFFAOYSA-N
2-methoxy-6-trifluoromethyl benzoyl chloride, 6-methoxy-2-trifluoromethyl benzoyl chloride, 2-methoxy-6-trifluoromethyl benzoyl chloride, jrd
15017534
2-methoxy-6-(trifluoromethyl)benzoyl chloride
COC1=CC=CC(=C1C(Cl)=O)C(F)(F)F
This item is not returnable. View return policy
681510-01-0
C9H6ClF3O2
238.59
MFCD09832320
HXPBLTYRTHTMFQ-UHFFFAOYSA-N
2-methoxy-6-trifluoromethyl benzoyl chloride, 6-methoxy-2-trifluoromethyl benzoyl chloride, 2-methoxy-6-trifluoromethyl benzoyl chloride, jrd
15017534
2-methoxy-6-(trifluoromethyl)benzoyl chloride
COC1=CC=CC(=C1C(Cl)=O)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.481
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 238.59
Percent Purity 97%
Chemical Name or Material 2-Methoxy-6-(trifluoromethyl)benzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'