Learn More
2-Methoxy-6-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™
Catalog Number p-7031211
Click to view available options
Quantity:
1 g
5 g
1017778-98-1
C9H7F3O2
204.15
MFCD09832324
LLBTYKHVRXDHKF-UHFFFAOYSA-N
2-methoxy-6-trifluoromethyl benzaldehyde, benzaldehyde, 2-methoxy-6-trifluoromethyl, 6-methoxy-2-trifluoromethyl benzaldehyde, 2-methoxy-6-trifluoromethyl benzaldehyde, jrd
20111744
2-methoxy-6-(trifluoromethyl)benzaldehyde
COC1=CC=CC(=C1C=O)C(F)(F)F
1017778-98-1
C9H7F3O2
204.15
MFCD09832324
LLBTYKHVRXDHKF-UHFFFAOYSA-N
2-methoxy-6-trifluoromethyl benzaldehyde, benzaldehyde, 2-methoxy-6-trifluoromethyl, 6-methoxy-2-trifluoromethyl benzaldehyde, 2-methoxy-6-trifluoromethyl benzaldehyde, jrd
20111744
2-methoxy-6-(trifluoromethyl)benzaldehyde
COC1=CC=CC(=C1C=O)C(F)(F)F
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Used as catalyst in diethylzinc addition to aldehydes.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY
Melting Point 46°C to 49°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 204.15
Percent Purity 97%
Chemical Name or Material 2-Methoxy-6-(trifluoromethyl)benzaldehyde