missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-6-(tri-n-butylstannyl)pyrazine, 95%, Thermo Scientific Chemicals

Catalog Number 15438117
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1105511-66-7
C17H32N2OSn
399.166
MFCD08275735
WWRXJELRWGUPKZ-UHFFFAOYSA-N
2-methoxy-6-tributylstannyl pyrazine, 2-tri-n-butylstannyl-6-methoxypyrazine, 2-methoxy-6-tributylstannanyl pyrazine, 2-methoxy-6-tri-n-butylstannyl pyrazine, 2-tributylstannyl-6-methoxypyrazine
16427103
tributyl-(6-methoxypyrazin-2-yl)stannane
CCCC[Sn](CCCC)(CCCC)C1=NC(=CN=C1)OC
This item is not returnable. View return policy
1105511-66-7
C17H32N2OSn
399.166
MFCD08275735
WWRXJELRWGUPKZ-UHFFFAOYSA-N
2-methoxy-6-tributylstannyl pyrazine, 2-tri-n-butylstannyl-6-methoxypyrazine, 2-methoxy-6-tributylstannanyl pyrazine, 2-methoxy-6-tri-n-butylstannyl pyrazine, 2-tributylstannyl-6-methoxypyrazine
16427103
tributyl-(6-methoxypyrazin-2-yl)stannane
CCCC[Sn](CCCC)(CCCC)C1=NC(=CN=C1)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN2788
Formula Weight 399.15
Percent Purity 95%
Chemical Name or Material 2-Methoxy-6-(tri-n-butylstannyl)pyrazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'