Få mere at vide
2-Methoxy-5-sulphamoylbenzamide, LoGiCal
Artikelnummer. 30463952
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463952 10 mg
1 options
Artikelnummer. 30463952 Leverandør LoGiCal Leverandørnr. LGCFOR0051.05
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methoxy-5-sulfamoylbenzamide
CAS 52395-25-2
Analyt- eller komponentnavne 2-Methoxy-5-sulfamoylbenzamide
Forsendelsestilstand Room Temperature
Molekylær formel C8 H10 N2 O4 S
Certificeringer/compliance ISO 9001
Synonym 2-Methoxy-5-sulphamoylbenzamide, Sulpiride Imp. E (EP), Sulpiride Impurity E
InChI formel InChI=1S/C8H10N2O4S/c1-14-7-3-2-5(15(10,12)13)4-6(7)8(9)11/h2-4H,1H3,(H2,9,11)(H2,10,12,13)
Løsningstype Neat
SMIL COc1ccc(cc1C(=O)N)S(=O)(=O)N
IUPAC navn 2-methoxy-5-sulfamoylbenzamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 230.24
Mængde 10 mg
Formel vægt 230.0361 g/mol