missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Methoxy-5-(pentafluorothio)phenylacetonitrile, 97%

Brand:  Alfa Aesar™ H33451.03

Additional Details : Weight : 0.00100kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15438475

  • 2251.00 DKK / 1g

Estimated Shipment
09-03-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1240257-27-5
273.221
UYISHVGSSKNHCB-UHFFFAOYSA-N
66523619
COC1=C(C=C(C=C1)S(F)(F)(F)(F)F)CC#N
C9H8F5NOS
MFCD16652489
2-methoxy-5-pentafluorosulfur phenylacetonitrile, 2-2-methoxy-5-pentafluoro-??-sulfanyl phenyl acetonitrile, 2-methoxy-5-pentafluorothio phenylacetonitrile
2-[2-methoxy-5-(pentafluoro-$l^{6}-sulfanyl)phenyl]acetonitrile

Specifications

2-Methoxy-5-(pentafluorothio)phenylacetonitrile
1240257-27-5
MFCD16652489
2-methoxy-5-pentafluorosulfur phenylacetonitrile, 2-2-methoxy-5-pentafluoro-??-sulfanyl phenyl acetonitrile, 2-methoxy-5-pentafluorothio phenylacetonitrile
COC1=C(C=C(C=C1)S(F)(F)(F)(F)F)CC#N
273.221
273.22
1g
C9H8F5NOS
UN3439
UYISHVGSSKNHCB-UHFFFAOYSA-N
2-[2-methoxy-5-(pentafluoro-$l^{6}-sulfanyl)phenyl]acetonitrile
66523619
97%
Videos
SDS Documents