missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-nitropyridine, 97%, Thermo Scientific™

Catalog Number p-157489
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
5446-92-4
C6H6N2O3
154.13
MFCD00006263
WUPLOZFIOAEYMG-UHFFFAOYSA-N
2-methoxy-5-nitro-pyridine, 5-nitro-2-methoxypyridine, 6-methoxy-3-nitropyridine, pyridine, 2-methoxy-5-nitro, methoxy 2--5-nitropyridine, 2-methoxy-5-nitropyridin, pubchem1207, acmc-1art3, 2-methoxy-5-nitro pyridine, 5-nitro-2-methoxy pyridine
79525
2-methoxy-5-nitropyridine
COC1=CC=C(C=N1)[N+]([O-])=O
This item is not returnable. View return policy
5446-92-4
C6H6N2O3
154.13
MFCD00006263
WUPLOZFIOAEYMG-UHFFFAOYSA-N
2-methoxy-5-nitro-pyridine, 5-nitro-2-methoxypyridine, 6-methoxy-3-nitropyridine, pyridine, 2-methoxy-5-nitro, methoxy 2--5-nitropyridine, 2-methoxy-5-nitropyridin, pubchem1207, acmc-1art3, 2-methoxy-5-nitro pyridine, 5-nitro-2-methoxy pyridine
79525
2-methoxy-5-nitropyridine
COC1=CC=C(C=N1)[N+]([O-])=O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 108.0°C to 111.0°C
Color Colorless to Yellow
Density 0.45g/mL
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3O(C5H3N)NO2
Quantity 25 g
Beilstein 21, II, 33
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chlorinated hydrocarbons,,insoluble in hexane
Formula Weight 154.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methoxy-5-nitropyridine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methoxy-5-nitropyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'