Få mere at vide
2-Methoxy-5-aminoformanilide, TRC
Artikelnummer. 30481268
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481268 250 mg
30466800 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30481268 Leverandør Toronto Research Chemicals Leverandørnr. TRCM260895250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 78939-34-1
Molekylær formel C8 H10 N2 O2
Molekylvægt (g/mol) 166.18
Synonym Formamide, N-(5-amino-2-methoxyphenyl)-, N-(5-Amino-2-methoxyphenyl)formamide, 2-Methoxy-5-aminoformanilide
IUPAC navn N-(5-amino-2-methoxyphenyl)formamide
SMIL COc1ccc(N)cc1NC=O
Tekniske data
Kemisk navn eller materiale 2-Methoxy-5-aminoformanilide
Mængde 250 mg
InChI formel InChI=1S/C8H10N2O2/c1-12-8-3-2-6(9)4-7(8)10-5-11/h2-5H,9H2,1H3,(H,10,11)
Formel vægt 166.0742