missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-(trifluoromethoxy)benzoic acid, 97%, Thermo Scientific™

Catalog Number p-7029106
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
848947-91-1
C9H7F3O4
236.146
MFCD04115966
GZSXIWDPTFEOFF-UHFFFAOYSA-N
2-methoxy-4-trifluoromethoxy benzoic acid, 2-methoxy-4-trifluoromethoxy-benzoic acid, benzoic acid, 2-methoxy-4-trifluoromethoxy
3258908
2-methoxy-4-(trifluoromethoxy)benzoic acid
COC1=C(C=CC(=C1)OC(F)(F)F)C(=O)O
This item is not returnable. View return policy
848947-91-1
C9H7F3O4
236.146
MFCD04115966
GZSXIWDPTFEOFF-UHFFFAOYSA-N
2-methoxy-4-trifluoromethoxy benzoic acid, 2-methoxy-4-trifluoromethoxy-benzoic acid, benzoic acid, 2-methoxy-4-trifluoromethoxy
3258908
2-methoxy-4-(trifluoromethoxy)benzoic acid
COC1=C(C=CC(=C1)OC(F)(F)F)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 87°C to 89°C
Quantity 1 g
Formula Weight 236.15
Percent Purity 97%
Chemical Name or Material 2-Methoxy-4-(trifluoromethoxy)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'