Få mere at vide
2-Methoxy-4-(prop-2-enyl)phenyl Acetate (Acetyleugenol), Mikromol™
Artikelnummer. 30049454
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30049454 100 mg
1 options
Artikelnummer. 30049454 Leverandør Mikromol Leverandørnr. MM3464.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Methoxy-4-(prop-2-enyl)phenyl Acetate (Acetyleugenol)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetyl Eugenol
CAS 93-28-7
Analyt- eller komponentnavne Acetyl Eugenol
Forsendelsestilstand Room Temperature
Molekylær formel C12 H14 O3
Certificeringer/compliance ISO 17025
Synonym Eugenol acetate (6CI), Phenol, 2-methoxy-4-(2-propenyl)-, acetate (9CI), Phenol, 4-allyl-2-methoxy-, acetate (7CI,8CI), 2-Methoxy-4-(2-propenyl)phenol acetate, 4-Allyl-2-methoxyphenyl acetate, Aceteugenol, Acetyleugenol, Eugenyl acetate, NSC 1242, O-Acetyleugenol, Phenol, 2-methoxy-4-(2-propen-1-yl)-, 1-acetate, Eugenol Imp. I (EP), Eugenol Impurity I
InChI formel InChI=1S/C12H14O3/c1-4-5-10-6-7-11(15-9(2)13)12(8-10)14-3/h4,6-8H,1,5H2,2-3H3
SMIL COc1cc(CC=C)ccc1OC(=O)C
IUPAC navn (2-methoxy-4-prop-2-enylphenyl) acetate
Molekylvægt (g/mol) 206.24
Mængde 100 mg
Formel vægt 206.0943 g/mol