Learn More
Alfa Aesar™ 2-Methoxy-4,6-bis(trifluoromethyl)benzonitrile, 97%
Brand: Alfa Aesar™ H31652.MD
Additional Details : Weight : 0.00025kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III
Quantity | 250mg |
---|
Chemical Identifiers
1092460-75-7 | |
269.146 | |
UEQPQBOSRAOLAK-UHFFFAOYSA-N | |
57364458 | |
COC1=CC(=CC(=C1C#N)C(F)(F)F)C(F)(F)F |
C10H5F6NO | |
MFCD11519321 | |
2-methoxy-4,6-bis trifluoromethyl benzonitrile, 2-methoxy-4,6-bis trifluoromethyl benzonitrile, jrd | |
2-methoxy-4,6-bis(trifluoromethyl)benzonitrile |
Specifications
2-Methoxy-4,6-bis(trifluoromethyl)benzonitrile | |
250mg | |
C10H5F6NO | |
UN3439 | |
UEQPQBOSRAOLAK-UHFFFAOYSA-N | |
2-methoxy-4,6-bis(trifluoromethyl)benzonitrile | |
57364458 | |
97% |
46°C to 48°C | |
1092460-75-7 | |
MFCD11519321 | |
2-methoxy-4,6-bis trifluoromethyl benzonitrile, 2-methoxy-4,6-bis trifluoromethyl benzonitrile, jrd | |
COC1=CC(=CC(=C1C#N)C(F)(F)F)C(F)(F)F | |
269.146 | |
269.14 |