Få mere at vide
2-Methoxy-4,5-dihydro-1H-imidazole, CDN
Artikelnummer. 30317161
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30317161 0.25 g -
30312612 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30317161 Leverandør CDN Leverandørnr. CDNX64910.25G
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4,5-Dihydro-2-methoxy-1 H-imidazole
CAS 28118-54-9
Molekylær formel C4 H8 N2 O
Mængde 0.25 g
Synonym 1 H-Imidazole, 4,5-dihydro-2-methoxy- (9 CI, ACI), 2-Imidazoline, 2-methoxy- (6 CI, 8 CI), 4,5-Dihydro-2-methoxy-1 H-imidazole (ACI), 2-Methoxy-2-imidazoline, 2-Methoxyimidazoline, 2-Methoxy-4,5-dihydro-1 H-imidazole, 2-Methoxy-4,5-dihydro-1 H-imidazole
InChI formel InChI=1 S/C4H8N2O/c1-7-4-5-2-3-6-4/h2-3H2,1H3,(H,5,6)
SMIL COC1=NCCN1
IUPAC navn 2-methoxy-4,5-dihydro-1 H-imidazole
Molekylvægt (g/mol) 100.119
Formel vægt 100.064 g/mol
Procent renhed min 98% chemical purity