Få mere at vide
2-Methoxy-4,5-dihydro-1H-imidazole-4,4,5,5-d4, CDN
Artikelnummer. 30316008
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30316008 0.05 g -
30310808 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316008 Leverandør CDN Leverandørnr. CDND61200.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4,5-Dihydro-2-methoxy-1 H-imidazole-4,5-d4
CAS 402788-68-5
Molekylær formel C4 2H4 H4 N2 O
Mængde 0.05 g
InChI formel InChI=1 S/C4H8N2O/c1-7-4-5-2-3-6-4/h2-3H2,1H3,(H,5,6)/i2D2,3D2
SMIL [2 H]C1([2 H])NC(=NC1([2 H])[2 H])OC
IUPAC navn 4,4,5,5-tetradeuterio-2-methoxy-1 H-imidazole
Molekylvægt (g/mol) 104.144
Formel vægt 104.089 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity